ω,3-Dibromoacetophenone 97%
Supplier: Apollo Scientific
Danger
Synonyms: 2-Bromo-1-(3-bromophenyl)ethanone, 2-Bromo-1-(3-bromophenyl)-1-ethanone, α,3-Dibromoacetophenone, 2,3'-Dibromoacetophenone, 3-Bromophenacyl bromide
OR5960-100G OR5960-25G OR5960-5G
APOSOR5960-100GEA 422 EUR
APOSOR5960-100G APOSOR5960-25G APOSOR5960-5G
ω,3-Dibromoacetophenone 97%
ω,3-Dibromoacetophenone
Formula: C₈H₆Br₂O
Melting Pt: 47…52 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00082698
CAS Number: 18523-22-3
UN: 3261
ADR: 8,II