Ethyl isothiocyanatoformate 98%
Supplier: Apollo Scientific
Danger
OR59819-100G OR59819-25G OR59819-5G
APOSOR59819-100GEA 271 EUR
APOSOR59819-100G APOSOR59819-25G APOSOR59819-5G
Ethyl isothiocyanatoformate 98%
Ethyl isothiocyanatoformate
Formula: C₄H₅NO₂S
Boiling Pt: 56 °C/18mmHg
Density: 1.112 g/cm³ (25 °C)
Flash Pt: 50 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004814
CAS Number: 16182-04-0
EINECS: 240-318-9
UN: 1992
ADR: 3+6.1,III