2-Thiophenemethanol 99%
Supplier: Apollo Scientific
Warning
Synonyms: Thiophen-2-methanol, Thiophen-2-ylmethanol, 2-Thienylmethanol
OR59889-100G OR59889-25G OR59889-500G
APOSOR59889-100GEA 97.3 EUR
APOSOR59889-100G APOSOR59889-25G APOSOR59889-500G
2-Thiophenemethanol 99%
2-Thiophenemethanol
Formula: C₅H₆OS
MW: 114.17 g/mol
Boiling Pt: 206…207 °C
Melting Pt: 0
Density: 1.205 g/cm³ (25 °C)
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005454
CAS Number: 636-72-6
EINECS: 211-264-3