3-Bromobenzoyl chloride
Supplier: Apollo Scientific
Danger
Synonyms: m-Bromobenzoyl chloride, 3-Bromobenzoylchloride
OR59972-25G OR59972-5G
APOSOR59972-25GEA 40.6 EUR
APOSOR59972-25G APOSOR59972-5G
3-Bromobenzoyl chloride
3-Bromobenzoyl chloride
Formula: C₇H₄BrClO
MW: 219.46 g/mol
Boiling Pt: 247…251 °C
Density: 1.662 g/cm³ (25 °C)
Flash Pt: 107 °C
Storage Temperature: Ambient
MDL Number: MFCD00000669
CAS Number: 1711-09-7
EINECS: 216-978-9
UN: 3265
ADR: 8 III