4-Iodobenzyl alcohol
Supplier: Apollo Scientific
Warning
Synonyms: (4-Iodophenyl)methanol
OR60044-5G OR60044-10G OR60044-25G
APOSOR60044-5GEA 65 EUR
APOSOR60044-5G APOSOR60044-10G APOSOR60044-25G
4-Iodobenzyl alcohol
4-Iodobenzyl alcohol
Formula: C₇H₇IO
Melting Pt: 72…75 °C
Storage Temperature: Ambient
MDL Number: MFCD01732720
CAS Number: 18282-51-4