1H-Indazole-7-carboxylic acid 97%
Supplier: Apollo Scientific
Warning
OR60064-1G OR60064-10G OR60064-5G
APOSOR60064-1GEA 29 EUR
APOSOR60064-1G APOSOR60064-10G APOSOR60064-5G
1H-Indazole-7-carboxylic acid 97%
1H-Indazole-7-carboxylic acid
Formula: C₈H₆N₂O₂
Melting Pt: 222…225 °C
Storage Temperature: Ambient
MDL Number: MFCD06804572
CAS Number: 677304-69-7