Imidazo[1,2-a]pyridine-6-carbonyl chloride hydrochloride
Supplier: Apollo Scientific
OR60211-1G OR60211-250MG
APOSOR60211-1GEA 216 EUR
APOSOR60211-1G APOSOR60211-250MG
Imidazo[1,2-a]pyridine-6-carbonyl chloride hydrochloride
Imidazo[1,2-a]pyridine-6-carbonyl chloride hydrochloride
Formula: C₈H₆Cl₂N₂O
MW: 217.049 g/mol
Melting Pt: 250…253 °C
Storage Temperature: Ambient
MDL Number: MFCD07772809
CAS Number: 859833-15-1