2-Amino-4-methoxynicotinonitrile
Supplier: Apollo Scientific
Warning
Synonyms: 2-Amino-4-methoxy-3-pyridinecarbonitrile
OR6055-1G OR6055-5G
APOSOR6055-1GEA 119 EUR
APOSOR6055-1G APOSOR6055-5G
2-Amino-4-methoxynicotinonitrile
2-Amino-4-methoxynicotinonitrile
Formula: C₇H₇N₃O
Melting Pt: 211 °C
Storage Temperature: Ambient
MDL Number: MFCD00975441
CAS Number: 98651-70-8