Trichloroacetamide
Supplier: Apollo Scientific
Warning
OR61119-100G OR61119-250G
APOSOR61119-100GEA 26.1 EUR
APOSOR61119-100G APOSOR61119-250G
Trichloroacetamide
Trichloroacetamide
Formula: C₂H₂Cl₃NO
Boiling Pt: 238…240 °C/760mmHg
Melting Pt: 139…141 °C
Storage Temperature: Ambient
MDL Number: MFCD00008009
CAS Number: 594-65-0
EINECS: 209-849-3