2-Methyl-10H-benzo[b]thieno[2,3-e][1,4]diazepin-4-amine hydrochloride
Supplier: Apollo Scientific
Warning
OR61175-1G OR61175-5G
APOSOR61175-1GEA 24.4 EUR
APOSOR61175-1G APOSOR61175-5G
2-Methyl-10H-benzo[b]thieno[2,3-e][1,4]diazepin-4-amine hydrochloride
2-Methyl-10H-benzo[b]thieno[2,3-e][1,4]diazepin-4-amine hydrochloride
Formula: C₁₂H₁₂ClN₃S
Melting Pt: 283…285 °C
Storage Temperature: Freezer
MDL Number: MFCD03411581
CAS Number: 138564-60-0