4-Tolualdehyde 96%
Supplier: Apollo Scientific
Warning
Synonyms: p-Tolualdehyde, p-Tolylaldehyde, p-Methylbenzaldehyd, 4-Methylbenzaldehyd, 4-Methylbenzaldehyde, 4-Tolylaldehyde
OR61250-100G OR61250-2.5KG OR61250-500G
APOSOR61250-100GEA 21.9 EUR
APOSOR61250-100G APOSOR61250-2.5KG APOSOR61250-500G
4-Tolualdehyde 96%
4-Tolualdehyde
Formula: C₈H₈O
MW: 120.15 g/mol
Boiling Pt: 204 °C
Melting Pt: –6 °C
Density: 1.018 g/cm³ (25 °C)
Flash Pt: 80 °C
Storage Temperature: Ambient
MDL Number: MFCD00006954
CAS Number: 104-87-0
EINECS: 203-246-9