2-Amino-6-methylpyrazine 97%
Supplier: Apollo Scientific
Warning
Synonyms: 6-Methylpyrazin-2-amine
OR6126-1G OR6126-250MG OR6126-5G
APOSOR6126-1GEA 66.7 EUR
APOSOR6126-1G APOSOR6126-250MG APOSOR6126-5G
2-Amino-6-methylpyrazine 97%
2-Amino-6-methylpyrazine
Formula: C₅H₇N₃
MW: 109.13 g/mol
Melting Pt: 121…128 °C
Storage Temperature: Ambient
MDL Number: MFCD08235193
CAS Number: 5521-56-2