5-Phenyl-1,3,4-oxadiazol-2-amine 97%
Supplier: Apollo Scientific
Warning
OR6135-1G OR6135-5G
APOSOR6135-1GEA 24.4 EUR
APOSOR6135-1G APOSOR6135-5G
5-Phenyl-1,3,4-oxadiazol-2-amine 97%
5-Phenyl-1,3,4-oxadiazol-2-amine
Formula: C₈H₇N₃O
Melting Pt: 237…242 °C
Storage Temperature: Ambient
MDL Number: MFCD00126383
CAS Number: 1612-76-6