2,5-Dichloronicotinonitrile 97%
Supplier: Apollo Scientific
Warning
OR61381-1G OR61381-5G
APOSOR61381-1GEA 23 EUR
APOSOR61381-1G APOSOR61381-5G
2,5-Dichloronicotinonitrile 97%
2,5-Dichloronicotinonitrile
Formula: C₆H₂Cl₂N₂
Melting Pt: 121…123 °C
Storage Temperature: Ambient
MDL Number: MFCD13185481
CAS Number: 126954-66-3