(±)-1-Boc-3-aminopyrrolidine 97%
Supplier: Apollo Scientific
Danger
Synonyms: 3-Aminopyrrolidine, N1-BOC protected, 1-Boc-3-Aminopyrrolidine
OR6139-25G OR6139-5G OR6139-1G
APOSOR6139-25GEA 112 EUR
APOSOR6139-25G APOSOR6139-5G APOSOR6139-1G
(±)-1-Boc-3-aminopyrrolidine 97%
(±)-1-Boc-3-aminopyrrolidine
Formula: C₉H₁₈N₂O₂
MW: 186.25 g/mol
Density: 1.022 g/cm³ (25 °C)
Flash Pt: 91 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01861220
CAS Number: 186550-13-0
UN: 2810
ADR: 6.1 III