(S)-Isopropyl-2-aminopropanoate hydrochloride
Supplier: Apollo Scientific
Warning
Synonyms: (S)-Isopropyl 2-aminopropanoate hydrochloride, (S)-2-Aminopropionic acid Isopropyl ester hydrochloride, L-Alanine Isopropyl ester hydrochloride, H-Ala-OiPr·HCl, Isopropyl (S)-2-Aminopropionate hydrochloride
OR61429-100G OR61429-25G
APOSOR61429-100GEA 42.2 EUR
APOSOR61429-100G APOSOR61429-25G
(S)-Isopropyl-2-aminopropanoate hydrochloride
(S)-Isopropyl-2-aminopropanoate hydrochloride
Formula: C₆H₁₃NO₂·HCl
MW: 167.6 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08059709
CAS Number: 62062-65-1