3-(4-Chlorophenyl)-1-methyl-1H-pyrazol-5-amine
Supplier: Apollo Scientific
Warning
Synonyms: 5-Amino-3-(4-chlorophenyl)-1-methyl-1H-pyrazole
OR6143-10G OR6143-1G OR6143-250MG
APOSOR6143-10GEA 241 EUR
APOSOR6143-10G APOSOR6143-1G APOSOR6143-250MG
3-(4-Chlorophenyl)-1-methyl-1H-pyrazol-5-amine
3-(4-Chlorophenyl)-1-methyl-1H-pyrazol-5-amine
Formula: C₁₀H₁₀ClN₃
Melting Pt: 150…152 °C
Storage Temperature: Ambient
MDL Number: MFCD00084872
CAS Number: 126417-82-1