3-Tolualdehyde 98%
Supplier: Apollo Scientific
Warning
Synonyms: m-Tolualdehyde, 3-Methylbenzaldehyde, m-Tolylaldehyde
OR61503-100G OR61503-500G OR61503-1KG
APOSOR61503-100GEA 46.4 EUR
APOSOR61503-100G APOSOR61503-500G APOSOR61503-1KG
3-Tolualdehyde 98%
3-Tolualdehyde
Formula: C₈H₈O
Boiling Pt: 198…200 °C
Density: 1.019 g/cm³ (25 °C)
Flash Pt: 78 °C
Storage Temperature: Ambient
MDL Number: MFCD00003374
CAS Number: 620-23-5
EINECS: 204-617-8