N-(4,6-Dimethylpyrimidin-2-yl)-N-methyl-3-aminobenzoic acid
Supplier: Apollo Scientific
Warning
OR6226-1G OR6226-250MG
APOSOR6226-1GEA 109 EUR
APOSOR6226-1G APOSOR6226-250MG
N-(4,6-Dimethylpyrimidin-2-yl)-N-methyl-3-aminobenzoic acid
N-(4,6-Dimethylpyrimidin-2-yl)-N-methyl-3-aminobenzoic acid
Formula: C₁₄H₁₅N₃O₂
Melting Pt: 168…170 °C
Storage Temperature: Ambient
MDL Number: MFCD00203266
CAS Number: 387350-52-9