3-Amino-4-hydroxybenzoic acid 97%
Supplier: Apollo Scientific
Warning
OR6238-100G OR6238-25G
APOSOR6238-100GEA 119 EUR
APOSOR6238-100G APOSOR6238-25G
3-Amino-4-hydroxybenzoic acid 97%
3-Amino-4-hydroxybenzoic acid
Formula: C₇H₇NO₃
Melting Pt: 198…208 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00007697
CAS Number: 1571-72-8
EINECS: 216-390-2