Salicylic acid 99%
Supplier: Apollo Scientific
Danger
OR6245-250G OR6245-500G OR6245-100G
APOSOR6245-250GEA 29 EUR
APOSOR6245-250G APOSOR6245-500G APOSOR6245-100G
Salicylic acid 99%
Salicylic acid
Formula: (HO)C₆H₄CO₂H
Boiling Pt: 211 °C/20mmHg
Melting Pt: 158…161 °C
Density: 1.44 g/cm³ (25 °C)
Flash Pt: 157 °C
Storage Temperature: Ambient
MDL Number: MFCD00002439
CAS Number: 69-72-7
EINECS: 200-712-3