2,3-Dimethylquinoxaline 97+%
Supplier: Apollo Scientific
Danger
OR6254-100G OR6254-25G
APOSOR6254-100GEA 171 EUR
APOSOR6254-100G APOSOR6254-25G
2,3-Dimethylquinoxaline 97+%
2,3-Dimethylquinoxaline
Formula: C₁₀H₁₀N₂
MW: 158.2 g/mol
Boiling Pt: 257…259 °C
Melting Pt: 104…108 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00006728
CAS Number: 2379-55-7
EINECS: 219-162-0
UN: 2811
ADR: 6.1 III