4'-tert-Butyl-2-chloroacetophenone 97%
Supplier: Apollo Scientific
Warning
Synonyms: 4-(tert-Butyl)phenacyl chloride
OR6263-1G OR6263-250MG OR6263-5G
APOSOR6263-1GEA 118 EUR
APOSOR6263-1G APOSOR6263-250MG APOSOR6263-5G
4'-tert-Butyl-2-chloroacetophenone 97%
4'-tert-Butyl-2-chloroacetophenone
Formula: C₉H₉ClO
Boiling Pt: 142…146 °C/1mmHg
Melting Pt: 36 °C
Storage Temperature: Ambient
MDL Number: MFCD00171621
CAS Number: 21886-62-4