2,5-Dimethyl-thiophene-3-carboxylic acid 97%
Supplier: Apollo Scientific
Warning
OR6267-1G OR6267-500MG
APOSOR6267-1GEA 379 EUR
APOSOR6267-1G APOSOR6267-500MG
2,5-Dimethyl-thiophene-3-carboxylic acid 97%
2,5-Dimethylthiophene-3-carboxylic acid
Formula: C₇H₈O₂S
MW: 156.2 g/mol
Melting Pt: 118 °C
Storage Temperature: Ambient
MDL Number: MFCD01860238
CAS Number: 26421-32-9