Methyl-1H-pyrrole-1-carboxylate 96%
Supplier: Apollo Scientific
Warning
OR6273-25G OR6273-5G
APOSOR6273-25GEA 698 EUR
APOSOR6273-25G APOSOR6273-5G
Methyl-1H-pyrrole-1-carboxylate 96%
Methyl-1H-pyrrole-1-carboxylate
Formula: C₆H₇NO₂
MW: 125.13 g/mol
Boiling Pt: 180 °C
Density: 1.113 g/cm³ (25 °C)
Flash Pt: 60 °C
Storage Temperature: Ambient
MDL Number: MFCD00126403
CAS Number: 4277-63-8
UN: 3272
ADR: 3 III