tert-Butyl-3-(2-methoxy-2-oxoethyl)piperazine-1-carboxylate 95%
Supplier: Apollo Scientific
Warning
Synonyms: 3-(2-Methoxy-2-oxoethyl)piperazine, N1-BOC protected
OR6400-1G OR6400-500MG OR6400-250MG
APOSOR6400-1GEA 459 EUR
APOSOR6400-1G APOSOR6400-500MG APOSOR6400-250MG
tert-Butyl-3-(2-methoxy-2-oxoethyl)piperazine-1-carboxylate 95%
tert-Butyl-3-(2-methoxy-2-oxoethyl)piperazine-1-carboxylate
Formula: C₁₂H₂₂N₂O₄
MW: 258.31 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01861213
CAS Number: 183742-33-8