3-Bromo-4-chlorotoluene

Supplier: Apollo Scientific
Warning

OR6447-100G OR6447-25G OR6447-5G
APOSOR6447-100GEA 190 EUR
APOSOR6447-100G APOSOR6447-25G APOSOR6447-5G
3-Bromo-4-chlorotoluene
3-Bromo-4-chlorotoluene
Formula: C₇H₆BrCl
MW: 205.48 g/mol
Boiling Pt: 112 °C/22mmHg
Density: 1.55 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00079710
CAS Number: 57310-39-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR