1,2-Dichloro-4-iodobenzene 99%
Supplier: Apollo Scientific
Warning
Synonyms: 3,4-Dichloroiodobenzene
OR6486-100G OR6486-250G OR6486-25G OR6486-500G
APOSOR6486-100GEA 49.3 EUR
APOSOR6486-100G APOSOR6486-250G APOSOR6486-25G APOSOR6486-500G
1,2-Dichloro-4-iodobenzene 99%
1,2-Dichloro-4-iodobenzene
Formula: C₆H₃Cl₂I
MW: 272.9 g/mol
Boiling Pt: 116…118 °C/12mmHg
Melting Pt: 27…29 °C
Storage Temperature: Ambient
MDL Number: MFCD00019014
CAS Number: 20555-91-3
EINECS: 243-874-0