Indoline 98%
Supplier: Apollo Scientific
Warning
Synonyms: 2,3-Dihydroindol
OR6520-250G OR6520-25G
APOSOR6520-250GEA 157 EUR
APOSOR6520-250G APOSOR6520-25G
Indoline 98%
Indoline
Formula: C₈H₉N
MW: 119.17 g/mol
Boiling Pt: 220…221 °C
Melting Pt: –21 °C
Density: 1.063 g/cm³ (25 °C)
Flash Pt: 92 °C
Storage Temperature: Ambient
MDL Number: MFCD00005705
CAS Number: 496-15-1
EINECS: 207-816-8