3-Cyclopentylpropionic acid
Supplier: Apollo Scientific
Warning
OR6533-100G OR6533-25G
APOSOR6533-100GEA 158 EUR
APOSOR6533-100G APOSOR6533-25G
3-Cyclopentylpropionic acid
3-Cyclopentylpropionic acid
Formula: C₈H₁₄O₂
Boiling Pt: 130…133 °C/12mmHg
Density: 0.996 g/cm³ (25 °C)
Flash Pt: 47 °C
Storage Temperature: Ambient
MDL Number: MFCD00001392
CAS Number: 140-77-2
EINECS: 205-433-0
UN: 1993
ADR: 3,III