Dibenzosuberenone 98%
Supplier: Apollo Scientific
Warning
OR6557-100G OR6557-25G
APOSOR6557-100GEA 68.6 EUR
APOSOR6557-100G APOSOR6557-25G
Dibenzosuberenone 98%
Dibenzosuberenone
Formula: C₁₅H₁₀O
Melting Pt: 87…88 °C
Storage Temperature: Ambient
MDL Number: MFCD00003588
CAS Number: 2222-33-5
EINECS: 218-737-3