1-Bromo-3-iodobenzene
Supplier: Apollo Scientific
Warning
Synonyms: 3-Bromoiodobenzene
OR6597-100G OR6597-1KG OR6597-250G
APOSOR6597-100GEA 36.4 EUR
APOSOR6597-100G APOSOR6597-1KG APOSOR6597-250G
1-Bromo-3-iodobenzene
1-Bromo-3-iodobenzene
Formula: C₆H₄BrI
Boiling Pt: 120 °C/18mmHg
Melting Pt: –10 to –9 °C
Density: 2.219 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00001043
CAS Number: 591-18-4
EINECS: 209-703-9