(R)-(+)-1,1'-Bi-2-naphthol 97%
Supplier: Apollo Scientific
Danger
Synonyms: (R)-[1,1'-Binaphthalene]-2,2'-diol, (R)-BINOL, (R)-(+)-1,1'-Binaphthyl-2,2'-Diol
OR6692-100G OR6692-250G OR6692-10G
APOSOR6692-100GEA 87 EUR
APOSOR6692-100G APOSOR6692-250G APOSOR6692-10G
(R)-(+)-1,1'-Bi-2-naphthol 97%
(R)-(+)-1,1'-Bi-2-naphthol
Formula: C₂₀H₁₄O₂
MW: 286.33 g/mol
Melting Pt: 208…211 °C
Storage Temperature: Ambient
MDL Number: MFCD00004068
CAS Number: 18531-94-7
UN: 2811
ADR: 6.1 III