2,3-Dimethylbenzaldehyde
Supplier: Apollo Scientific
Warning
OR6703-100G OR6703-25G
APOSOR6703-100GEA 221 EUR
APOSOR6703-100G APOSOR6703-25G
2,3-Dimethylbenzaldehyde
2,3-Dimethylbenzaldehyde
Formula: C₉H₁₀O
MW: 134.18 g/mol
Boiling Pt: 86…88 °C/10mmHg
Density: 1.029 g/cm³ (25 °C)
Flash Pt: 101 °C
Storage Temperature: Ambient
MDL Number: MFCD00798004
CAS Number: 5779-93-1