2-Pyrid-3-ylbenzoic acid 97%
Supplier: Apollo Scientific
Warning
Synonyms: 2-(Pyridin-3-yl)benzoic acid
OR6788-1G OR6788-250MG
APOSOR6788-1GEA 261 EUR
APOSOR6788-1G APOSOR6788-250MG
2-Pyrid-3-ylbenzoic acid 97%
2-Pyrid-3-ylbenzoic acid
Formula: C₁₂H₉NO₂
Melting Pt: 187.5…188.5 °C
Storage Temperature: Ambient
MDL Number: MFCD03426511
CAS Number: 134363-45-4