3-Chlorosalicylaldehyde 98%

Supplier: Apollo Scientific
Warning

Synonyms: 3-Chloro-2-hydroxybenzaldehyde, 2-Hydroxy-3-chlorobenzaldehyde

OR6968-1G OR6968-500MG
APOSOR6968-1GEA 69.6 EUR
APOSOR6968-1G APOSOR6968-500MG
3-Chlorosalicylaldehyde 98%
3-Chlorosalicylaldehyde
Formula: C₇H₅ClO₂
Melting Pt: 52…56 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD04973581
CAS Number: 1927-94-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR