6,6'-Dimethyl-2,2'-bipyridyl 98%
Supplier: Apollo Scientific
Warning
Synonyms: 6,6'-Dimethyl-2,2'-bipyridine
OR70013-5G OR70013-1G
APOSOR70013-5GEA 37.7 EUR
APOSOR70013-5G APOSOR70013-1G
6,6'-Dimethyl-2,2'-bipyridyl 98%
6,6'-Dimethyl-2,2'-bipyridyl
Formula: C₁₂H₁₂N₂
MW: 184.2371 g/mol
Boiling Pt: 286 °C
Melting Pt: 88 °C
Storage Temperature: Ambient
MDL Number: MFCD00059776
CAS Number: 4411-80-7