4,4'-Di-tert-butyl-2,2'-bipyridine 98%
Supplier: Apollo Scientific
Warning
Synonyms: 4,4'-Di-tert-butyl-2,2'-bipyridyl, 4,4'-Di-tert-butyl-2,2'-bipyridine
OR70014-1G OR70014-5G
APOSOR70014-1GEA 21.9 EUR
APOSOR70014-1G APOSOR70014-5G
4,4'-Di-tert-butyl-2,2'-bipyridine 98%
4,4'-Di-tert-butyl-2,2'-bipyridine
Formula: C₁₈H₂₄N₂
MW: 268.4000 g/mol
Melting Pt: 159…161 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01863731
CAS Number: 72914-19-3