2-Acetyl-1-methyl-1H-pyrrole
Supplier: Apollo Scientific
Warning
Synonyms: 1-(1-Methyl-1H-pyrrol-2-yl)ethanone, 2-Acetyl-1-methylpyrrole
OR7070-100G OR7070-25G
APOSOR7070-100GEA 88.6 EUR
APOSOR7070-100G APOSOR7070-25G
2-Acetyl-1-methyl-1H-pyrrole
2-Acetyl-1-methyl-1H-pyrrole
Formula: C₇H₉NO
Boiling Pt: 200…202 °C
Density: 1.04 g/cm³ (25 °C)
Flash Pt: 68 °C
Storage Temperature: Ambient
MDL Number: MFCD00003089
CAS Number: 932-16-1