Cyclohexylacetic acid 98%
Supplier: Apollo Scientific
Warning
Synonyms: Cyclohexaneacetic acid
OR7120-25G OR7120-5G OR7120-1G
APOSOR7120-25GEA 161 EUR
APOSOR7120-25G APOSOR7120-5G APOSOR7120-1G
Cyclohexylacetic acid 98%
Cyclohexylacetic acid
Formula: C₈H₁₄O₂
MW: 142.2 g/mol
Boiling Pt: 242…244 °C
Melting Pt: 28…30 °C
Density: 1.007 g/cm³ (25 °C)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00001518
CAS Number: 5292-21-7
EINECS: 226-132-0