9-Fluorenylmethanol
Supplier: Apollo Scientific
Warning
Synonyms: (Fluoren-9-yl)methanol, 9-Fluorenmethanol, 9-Fluorenemethanol
OR7130-100G OR7130-1KG OR7130-25G
APOSOR7130-100GEA 24.8 EUR
APOSOR7130-100G APOSOR7130-1KG APOSOR7130-25G
9-Fluorenylmethanol
9-Fluorenylmethanol
Formula: C₁₄H₁₂O
Melting Pt: 102…107 °C
Storage Temperature: Ambient
MDL Number: MFCD00001139
CAS Number: 24324-17-2
EINECS: 246-167-5