2-Iodosobenzoic acid 97%
Supplier: Apollo Scientific
Warning
OR7133-1G OR7133-5G
APOSOR7133-1GEA 94.5 EUR
APOSOR7133-1G APOSOR7133-5G
2-Iodosobenzoic acid 97%
2-Iodosobenzoic acid
Formula: C₇H₅IO₃
Melting Pt: 230 °C (dec.)
Storage Temperature: Freezer
MDL Number: MFCD00002401
CAS Number: 304-91-6
EINECS: 206-159-4