1-Methylcyclohexene
Supplier: Apollo Scientific
Danger
OR7135-100G OR7135-25G
APOSOR7135-100GEA 317 EUR
APOSOR7135-100G APOSOR7135-25G
1-Methylcyclohexene
1-Methylcyclohexene
Formula: C₇H₁₂
Boiling Pt: 110…111 °C
Density: 0.811 g/cm³ (25 °C)
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00001548
CAS Number: 591-49-1
UN: 3295
ADR: 3,II