4-Propylphenylboronic acid
Supplier: Apollo Scientific
Warning
OR7257-1G OR7257-5G OR7257-10G
APOSOR7257-1GEA 14.7 EUR
APOSOR7257-1G APOSOR7257-5G APOSOR7257-10G
4-Propylphenylboronic acid
4-Propylphenylboronic acid
Formula: C₉H₁₃BO₂
Melting Pt: 89…97 °C
Storage Temperature: Ambient
MDL Number: MFCD00859261
CAS Number: 134150-01-9