2,3-Dimethylthiophene
Supplier: Apollo Scientific
Warning
OR7265-1G OR7265-5G
APOSOR7265-1GEA 26.1 EUR
APOSOR7265-1G APOSOR7265-5G
2,3-Dimethylthiophene
2,3-Dimethylthiophene
Formula: C₆H₈S
MW: 112.2 g/mol
Boiling Pt: 141…142 °C
Melting Pt: –49 °C
Density: 1.0021
Flash Pt: 26 °C
Storage Temperature: Ambient
MDL Number: MFCD00130081
CAS Number: 632-16-6
EINECS: 211-170-2
UN: 1993
ADR: 3 III