4-Vinylbenzoic acid 97%
Supplier: Apollo Scientific
Warning
OR7329-25G OR7329-100G OR7329-10G
APOSOR7329-25GEA 109 EUR
APOSOR7329-25G APOSOR7329-100G APOSOR7329-10G
4-Vinylbenzoic acid 97%
4-Vinylbenzoic acid
Formula: C₉H₈O₂
Melting Pt: 142…144 °C
Density: 1.3 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00002569
CAS Number: 1075-49-6
EINECS: 214-053-4