2'-Methoxybiphenyl-4-carbaldehyde
Supplier: Apollo Scientific
Warning
Synonyms: 2'-Methoxy-[1,1'-biphenyl]-4-carbaldehyde, 2'-Methoxy-[1,1'-biphenyl]-4-carboxaldehyde
OR7454-1G OR7454-5G
APOSOR7454-1GEA 23.2 EUR
APOSOR7454-1G APOSOR7454-5G
2'-Methoxybiphenyl-4-carbaldehyde
2'-Methoxybiphenyl-4-carbaldehyde
Formula: C₁₄H₁₂O₂
Melting Pt: 54…55 °C
Storage Temperature: Ambient
MDL Number: MFCD01631913
CAS Number: 421553-62-0