(2-Chlorophenyl)acetylene 98%
Supplier: Apollo Scientific
Warning
Synonyms: 1-Chloro-2-ethynylbenzene
OR7551T-25G OR7551T-5G OR7551T-100G
APOSOR7551T-25GEA 186 EUR
APOSOR7551T-25G APOSOR7551T-5G APOSOR7551T-100G
(2-Chlorophenyl)acetylene 98%
(2-Chlorophenyl)acetylene
Formula: C₈H₅Cl
Boiling Pt: 82 °C/12mmHg
Density: 1.125 g/cm³ (25 °C)
Flash Pt: 70 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00269951
CAS Number: 873-31-4