Cyclooctanol
Supplier: Apollo Scientific
Warning
OR7566-25G OR7566-100G
APOSOR7566-25GEA 154 EUR
APOSOR7566-25G APOSOR7566-100G
Cyclooctanol
Cyclooctanol
Formula: C₈H₁₆O
Boiling Pt: 97…98 °C/11mmHg
Melting Pt: 14…15 °C
Density: 0.97 g/cm³ (25 °C)
Flash Pt: 86 °C
Storage Temperature: Ambient
MDL Number: MFCD00001744
CAS Number: 696-71-9
EINECS: 211-800-6