5-Nitro-o-toluonitrile
Supplier: Apollo Scientific
Warning
Synonyms: 2-Methyl-5-nitrobenzonitrile
OR7738-25G OR7738-5G OR7738-100G
APOSOR7738-25GEA 29 EUR
APOSOR7738-25G APOSOR7738-5G APOSOR7738-100G
5-Nitro-o-toluonitrile
5-Nitro-o-toluonitrile
Formula: C₈H₆N₂O₂
MW: 162.15
Boiling Pt: 174
Melting Pt: 103…108 °C
Storage Temperature: Ambient
MDL Number: MFCD00017040
CAS Number: 939-83-3